Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scaf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139094.1 | NP_620794.1 | 2 | 423 | ||
XM_006227858.2 | XP_006227920.1 | 3 | 350 | |||
GenBank mRNA | U49055.1 | - | 2 | 423 |