Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Dmgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139102.2 | NP_620802.2 | 3 | 299 | ||
XM_008760629.1 | XP_008758851.1 | 3 | 314 | |||
XM_008760630.1 | XP_008758852.1 | 3 | 314 | |||
XM_008760631.1 | XP_008758853.1 | 3 | 314 | |||
XM_008760632.1 | XP_008758854.1 | 3 | 314 | |||
GenBank mRNA | BC085697.1 | - | 3 | 299 | 7108858 | |
X55995.1 | - | 3 | 267 |