Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Crhbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139183.2 | NP_631922.1 | 3, 4 | 459 | ||
GenBank mRNA | X58023.1 | - | 3, 4 | 458 |