Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpsm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144745.2 | NP_653346.2 | 2 | 306 | ||
XM_006233670.2 | XP_006233732.1 | 2 | 410 | |||
GenBank mRNA | AF107723.1 | - | 1 | 430 | ||
CB743977.1 | - | 2 | 354 |