Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ddx46 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139098.1 | NP_620798.1 | 15 | 2018 | ||
XM_006253601.2 | XP_006253663.1 | 15 | 2048 | |||
GenBank mRNA | BC107590.1 | - | 15 | 1884 | 7461248 | |
U25746.1 | - | 15 | 2018 |