Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tpcn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139332.3 | NP_647548.2 | 22 | 2131 | ||
GenBank mRNA | AB018253.2 | - | 21 | 2134 | ||
BC062072.1 | - | 21 | 2131 | 5599545 |