Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Zfp180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001271280.1 | NP_001258209.1 | 5 | 1153 | ||
NM_144757.3 | NP_653358.2 | 5 | 1071 | |||
XM_006228386.2 | XP_006228448.1 | 5 | 1104 | |||
XM_006228387.2 | XP_006228449.1 | 5 | 1155 | |||
XM_008758890.1 | XP_008757112.1 | 623 | ||||
GenBank mRNA | U41164.1 | - | 5 | 1071 |