Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Optn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145081.3 | NP_659549.2 | 2 | 358 | ||
XM_006254262.2 | XP_006254324.1 | 3 | 516 | |||
XM_006254263.2 | XP_006254325.1 | 3 | 515 | |||
XM_006254264.2 | XP_006254326.1 | 3 | 515 | |||
XM_008771852.1 | XP_008770074.1 | 3 | 516 | |||
XM_008771853.1 | XP_008770075.1 | 3 | 515 | |||
GenBank mRNA | AB069907.3 | - | 2 | 358 | ||
AB222074.1 | - | 2 | 312 | |||
BC086976.1 | - | 3 | 444 | 7111026 | ||
FQ223875.1 | - | 3 | 515 |