Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 1, 2 | 193 | ||
NM_018427.4 | NP_060897.3 | 1, 2 | 193 | |||
XM_005255376.1 | XP_005255433.1 | 1, 2 | 102 | |||
XM_005255377.4 | XP_005255434.1 | 1, 2 | 141 | |||
XM_011522541.1 | XP_011520843.1 | 1, 2 | 102 | |||
GenBank mRNA | AF227156.1 | - | 1, 2 | 159 | ||
AJ272050.1 | - | 1, 2 | 113 | |||
AK225662.1 | - | 1, 2 | 169 | |||
AK300253.1 | - | 1, 2 | 102 | |||
AK314769.1 | - | 1, 2 | 107 | |||
AK316520.1 | - | 1, 2 | 107 | |||
BC036182.1 | - | 1, 2 | 108 | |||
BC104660.1 | - | 1, 2 | 115 | |||
BC132688.1 | - | 1, 2 | 110 | 40146710 | ||
BC132690.1 | - | 1, 2 | 110 | 40146712 | ||
DC376629.1 | - | 1, 2 | 194 |
Gene Symbol: RRN3P2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_003369.2 | 1, 2 | 282 | |||
GenBank mRNA | AK225670.1 | - | 1, 2 | 130 | ||
AK298442.1 | - | 1, 2 | 105 | |||
AK301253.1 | - | 1, 2 | 102 | |||
DC420905.1 | - | 1, 2 | 305 |