Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_173838.1 | NP_776210.1 | 1 | 611 | ||
GenBank mRNA | AB052909.1 | - | 1 | 611 |
Set Membership: |
![]() |