Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lzts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172022.2 | NP_742019.2 | 4 | 1925 | ||
XM_006235009.2 | XP_006235071.1 | 6 | 3154 | |||
XM_006235010.2 | XP_006235072.1 | 6 | 3906 | |||
XM_006235011.2 | XP_006235073.1 | 5 | 3106 | |||
XM_006235014.2 | XP_006235076.1 | 5 | 1976 | |||
XM_008762147.1 | XP_008760369.1 | 6 | 2943 | |||
XM_008762148.1 | XP_008760370.1 | 5 | 2894 | |||
XM_008762149.1 | XP_008760371.1 | 6 | 2881 | |||
XM_008762150.1 | XP_008760372.1 | 5 | 2832 | |||
XM_008762151.1 | XP_008760373.1 | 4 | 1967 | |||
GenBank mRNA | AJ278801.2 | - | 4 | 1925 |