Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Impa2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172224.1 | NP_757378.1 | 6 | 699 | ||
XM_006254856.2 | XP_006254918.1 | 6 | 4018 | |||
GenBank mRNA | AY160191.1 | - | 6 | 699 | ||
BC083544.1 | - | 6 | 673 | 7191219 |