Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Arfgef2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181083.2 | NP_851597.2 | 4 | 358 | ||
XM_006235640.2 | XP_006235702.1 | 4 | 543 | |||
GenBank mRNA | AY255526.2 | - | 4 | 358 |