Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CYSLTR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001308465.1 | NP_001295394.1 | 6 | 795 | ||
NM_001308467.1 | NP_001295396.1 | 6 | 772 | |||
NM_001308468.1 | NP_001295397.1 | 4 | 593 | |||
NM_001308469.1 | NP_001295398.1 | 4 | 570 | |||
NM_001308470.1 | NP_001295399.1 | 3 | 503 | |||
NM_001308471.1 | NP_001295400.1 | 2 | 402 | |||
NM_001308476.1 | NP_001295405.1 | 5 | 666 | |||
NM_020377.3 | NP_065110.1 | 5 | 643 | |||
GenBank mRNA | AB038269.1 | - | 1 | 357 | ||
AB041947.1 | - | 1 | 428 | |||
AK291739.1 | - | 5 | 643 | |||
AY853711.1 | - | 4 | 322 | |||
BC069160.1 | - | 1 | 97 | 7216884 | ||
BC096832.1 | - | 1 | 210 | 6971946 |
Set Membership: |
G-protein coupled receptor |