Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cd59 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012925.1 | NP_037057.1 | 3 | 171 | ||
GenBank mRNA | BC063176.1 | - | 3 | 165 | 6891456 | |
U48255.1 | - | 3 | 171 |