Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Twist2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021691.2 | NP_067723.1 | 1 | 115 | ||
GenBank mRNA | AI029397.1 | - | 1 | 183 | ||
Y08139.1 | - | 1 | 115 |