Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cela2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012553.2 | NP_036685.1 | 3 | 176 | ||
GenBank mRNA | FQ226773.1 | - | 3 | 178 | ||
FQ227652.1 | - | 3 | 178 | |||
FQ227675.1 | - | 3 | 178 | |||
FQ228000.1 | - | 3 | 178 | |||
FQ228239.1 | - | 2 | 59 | |||
V01233.1 | - | 3 | 176 |