Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nppa | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012612.2 | NP_036744.1 | 2 | 228 | ||
GenBank mRNA | BC158590.1 | - | 2 | 218 | 9036379 | |
CB724799.1 | - | 2 | 355 | |||
M15868.1 | - | 1 | 42 | |||
X00665.1 | - | 2 | 220 | |||
X01118.1 | - | 2 | 209 |