Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PVALB SIRNA Assays ›
Gene Symbol:
Pvalb
Gene Name:
parvalbumin
Gene Aliases:
PALB1, Pva
Chromosome Location:
Chr.7: 119428652 - 119443656 on Build Rnor_6.0
UniGene ID:
Rn.2005
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
59326
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Pvalb
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_022499.2
NP_071944.1
2
87
XM_006241929.2
XP_006241991.1
3
160
GenBank mRNA
BC126090.1
-
2
87
7321065
FQ212120.1
-
3
150
FQ214597.1
-
3
148
FQ214667.1
-
3
149
FQ214909.1
-
3
148
FQ215072.1
-
3
148
FQ215172.1
-
3
148
FQ215250.1
-
3
149
FQ215353.1
-
3
148
FQ215367.1
-
3
148
FQ215388.1
-
3
148
FQ215472.1
-
3
148
FQ215487.1
-
3
148
FQ215608.1
-
3
149
FQ215681.1
-
3
148
FQ216147.1
-
3
148
FQ216186.1
-
3
148
FQ216201.1
-
3
149
FQ216338.1
-
3
149
FQ216339.1
-
3
148
FQ216448.1
-
3
148
FQ216459.1
-
3
148
FQ216533.1
-
3
148
FQ216659.1
-
3
148
FQ216789.1
-
3
148
FQ216830.1
-
3
148
FQ216843.1
-
3
148
FQ216867.1
-
3
148
FQ216872.1
-
3
148
FQ216903.1
-
3
150
FQ216906.1
-
3
145
FQ216960.1
-
3
148
FQ216981.1
-
3
148
FQ217053.1
-
3
148
FQ217064.1
-
3
148
FQ217099.1
-
3
148
FQ217132.1
-
2
149
FQ217209.1
-
3
147
FQ217210.1
-
3
148
FQ217278.1
-
3
148
FQ217298.1
-
3
149
FQ217358.1
-
3
148
FQ217373.1
-
3
149
FQ217384.1
-
3
148
FQ217418.1
-
3
149
FQ217424.1
-
3
148
FQ217446.1
-
2
116
FQ217489.1
-
3
148
FQ217527.1
-
3
148
FQ217562.1
-
3
148
FQ217576.1
-
3
148
FQ217645.1
-
3
147
FQ217652.1
-
3
149
FQ217677.1
-
3
149
FQ217743.1
-
3
147
FQ217749.1
-
3
149
FQ217760.1
-
3
149
FQ217770.1
-
3
148
FQ217787.1
-
3
149
FQ217845.1
-
3
148
FQ217858.1
-
3
148
FQ217860.1
-
3
149
FQ217861.1
-
3
148
FQ217880.1
-
3
150
FQ217886.1
-
3
148
FQ217934.1
-
2
90
FQ217966.1
-
3
148
FQ217976.1
-
3
148
FQ217991.1
-
3
148
FQ218022.1
-
3
148
FQ218035.1
-
3
148
FQ218057.1
-
3
148
FQ223582.1
-
3
148
FQ223596.1
-
3
148
FQ223603.1
-
3
149
FQ223630.1
-
3
145
FQ223664.1
-
3
149
FQ223695.1
-
3
148
FQ223712.1
-
3
149
FQ223798.1
-
3
150
FQ223861.1
-
3
148
FQ223863.1
-
3
148
FQ223909.1
-
3
148
FQ223941.1
-
3
148
FQ223961.1
-
3
148
FQ223988.1
-
3
147
FQ223989.1
-
3
148
FQ223990.1
-
3
148
FQ224014.1
-
3
148
FQ224025.1
-
3
149
FQ224028.1
-
3
148
FQ224037.1
-
3
148
FQ224053.1
-
3
148
FQ224059.1
-
3
148
FQ224083.1
-
3
148
FQ224113.1
-
3
148
FQ224136.1
-
3
148
FQ224173.1
-
3
148
FQ224181.1
-
3
148
FQ224187.1
-
3
148
FQ224242.1
-
3
149
FQ224248.1
-
3
146
FQ224294.1
-
3
148
FQ224303.1
-
3
149
FQ224322.1
-
3
148
FQ224356.1
-
3
148
FQ224363.1
-
3
148
FQ224417.1
-
3
148
FQ224430.1
-
3
148
FQ224435.1
-
3
146
FQ224463.1
-
3
149
FQ224483.1
-
3
148
FQ224485.1
-
3
148
FQ224488.1
-
3
149
FQ224510.1
-
3
148
FQ224541.1
-
3
148
FQ224580.1
-
3
148
FQ224584.1
-
3
149
FQ224600.1
-
3
148
FQ224613.1
-
3
148
FQ224634.1
-
3
148
FQ224643.1
-
3
148
FQ224673.1
-
3
149
FQ224675.1
-
3
148
FQ224723.1
-
3
150
FQ224767.1
-
3
148
FQ224773.1
-
3
150
FQ224778.1
-
3
148
FQ224913.1
-
3
148
M12725.1
-
3
147
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_013645, BC027424, X54613, S75909, S75910, and others.
Panther Classification:
Molecular Function -
calmodulin-related
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytosolic calcium ion concentration
calcium ion binding
protein homodimerization activity
protein heterodimerization activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0