Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gng7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024138.1 | NP_077052.1 | 2 | 312 | ||
GenBank mRNA | DQ120497.1 | - | 1 | 70 | ||
DQ120498.1 | - | 1 | 70 | |||
FQ211653.1 | - | 292 | ||||
L23219.1 | - | 2 | 312 |