Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: P2ry6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_057124.2 | NP_476465.1 | 3 | 317 | ||
XM_006229711.2 | XP_006229773.1 | 2 | 401 | |||
XM_006229712.2 | XP_006229774.1 | 3 | 1753 | |||
XM_006229713.2 | XP_006229775.1 | 3 | 470 | |||
XM_006229714.2 | XP_006229776.1 | 3 | 1417 | |||
XM_006229715.2 | XP_006229777.1 | 4 | 1900 | |||
XM_006229716.2 | XP_006229778.1 | 3 | 1138 | |||
XM_006229717.2 | XP_006229779.1 | 3 | 3708 | |||
GenBank mRNA | BC072520.1 | - | 3 | 317 | 7105972 | |
D63665.1 | - | 4 | 541 |
Set Membership: |
G-protein coupled receptor |