Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Oaz1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001297557.1 | NP_001284486.1 | 1, 2 | 233 | ||
NM_139081.2 | NP_620781.1 | 1, 2 | 233 | |||
GenBank mRNA | D10706.1 | - | 1, 2 | 205 | ||
FQ211375.1 | - | 1, 2 | 233 | |||
FQ213504.1 | - | 1, 2 | 233 | |||
FQ213794.1 | - | 1, 2 | 233 | |||
FQ214019.1 | - | 1, 2 | 228 | |||
FQ214051.1 | - | 1, 2 | 228 | |||
FQ214373.1 | - | 1, 2 | 228 | |||
FQ214458.1 | - | 1, 2 | 228 | |||
FQ216817.1 | - | 1, 2 | 233 | |||
FQ218486.1 | - | 1, 2 | 228 | |||
FQ218990.1 | - | 1, 2 | 228 | |||
FQ220491.1 | - | 1, 2 | 228 | |||
FQ220580.1 | - | 1, 2 | 233 | |||
FQ225116.1 | - | 1, 2 | 228 | |||
FQ225909.1 | - | 1, 2 | 228 | |||
FQ225949.1 | - | 1, 2 | 231 | |||
FQ229954.1 | - | 1, 2 | 228 | |||
FQ230452.1 | - | 1, 2 | 228 | |||
FQ231088.1 | - | 1, 2 | 228 | |||
FQ232667.1 | - | 1, 2 | 228 | |||
FQ232997.1 | - | 1, 2 | 228 | |||
FQ233723.1 | - | 1, 2 | 228 |