Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ROCK2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321643.1 | NP_001308572.1 | 19 | 2227 | ||
NM_004850.4 | NP_004841.2 | 19 | 2683 | |||
XM_005246190.3 | XP_005246247.1 | 19 | 2717 | |||
XM_011510417.2 | XP_011508719.1 | 19 | 2182 | |||
XM_017005378.1 | XP_016860867.1 | 19 | 2714 | |||
XM_017005379.1 | XP_016860868.1 | 19 | 2112 | |||
GenBank mRNA | AB014519.1 | - | 19 | 2688 | ||
AK299296.1 | - | 5 | 690 | |||
AK310358.1 | - | 15 | 1978 | |||
BC111801.1 | - | 6 | 790 | 40069866 | ||
D87931.1 | - | 19 | 2234 |