Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ucp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019354.3 | NP_062227.2 | 3 | 463 | ||
GenBank mRNA | AB005143.1 | - | 1 | 90 | ||
AB006613.1 | - | 1 | 90 | |||
AB010743.1 | - | 3 | 434 | |||
AF039033.1 | - | 1 | 90 | |||
BC062230.1 | - | 3 | 435 | 6920447 | ||
FQ227097.1 | - | 3 | 463 |