Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptafr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_053321.2 | NP_445773.1 | 2 | 259 | ||
XM_006239063.2 | XP_006239125.1 | 2 | 584 | |||
XM_006239064.2 | XP_006239126.1 | 2 | 870 | |||
GenBank mRNA | BC072491.1 | - | 2 | 259 | 7097307 | |
U04740.1 | - | 2 | 280 |
Set Membership: |
![]() |