Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Sostdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153737.1 | NP_714959.1 | 1 | 171 | ||
GenBank mRNA | AF411056.1 | - | 1 | 171 | ||
BC081710.1 | - | 1 | 145 | 7191839 | ||
FQ213516.1 | - | 1 | 161 |