Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007413.4 | NP_031439.2 | 1, 2 | 461 | ||
GenBank mRNA | AK047002.1 | - | 1, 2 | 462 | ||
AK077289.1 | - | 1, 2 | 235 | |||
AK080759.1 | - | 1, 2 | 456 | |||
BC116415.1 | - | 1, 2 | 419 | 40103279 | ||
BC116416.1 | - | 1, 2 | 419 | 40103280 | ||
U05673.1 | - | 1, 2 | 376 |
Set Membership: |
G-protein coupled receptor |