Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Alpl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001287172.1 | NP_001274101.1 | 6 | 882 | ||
NM_001287176.1 | NP_001274105.1 | 6 | 1005 | |||
NM_007431.3 | NP_031457.2 | 6 | 857 | |||
GenBank mRNA | AB473959.1 | - | 5 | 621 | ||
AK076459.1 | - | 6 | 792 | |||
AK161780.1 | - | 6 | 793 | |||
AK164002.1 | - | 6 | 850 | |||
AK166732.1 | - | 6 | 791 | |||
AK167023.1 | - | 6 | 792 | |||
AK167481.1 | - | 6 | 792 | |||
BC065175.1 | - | 6 | 780 | 6807509 | ||
J02980.1 | - | 6 | 796 | |||
X13409.1 | - | 6 | 740 |