Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aatk | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001198785.1 | NP_001185714.1 | 8 | 1037 | ||
NM_001198787.1 | NP_001185716.1 | 8 | 809 | |||
NM_007377.4 | NP_031403.2 | 8 | 1320 | |||
GenBank mRNA | AB093253.2 | - | 8 | 1320 | ||
AB288871.1 | - | 8 | 939 | |||
AF011908.1 | - | 8 | 776 | |||
AK147239.1 | - | 5 | 830 | |||
AK155100.1 | - | 8 | 811 | |||
AY236858.1 | - | 8 | 815 | |||
AY236859.1 | - | 8 | 842 | |||
BC042837.1 | - | 8 | 1342 | |||
BC052323.1 | - | 8 | 1342 | |||
BC080846.1 | - | 8 | 891 | 6856191 |