Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Adora2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007413.4 | NP_031439.2 | 2 | 469 | ||
GenBank mRNA | AK047002.1 | - | 2 | 470 | ||
AK077289.1 | - | 2 | 243 | |||
AK080759.1 | - | 2 | 464 | |||
BC116415.1 | - | 2 | 427 | 40103279 | ||
BC116416.1 | - | 2 | 427 | 40103280 | ||
U05673.1 | - | 2 | 384 |
Set Membership: |
G-protein coupled receptor |