Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PLXDC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020405.4 | NP_065138.2 | 6 | 868 | ||
GenBank mRNA | AF279144.2 | - | 6 | 738 | ||
AF378753.1 | - | 6 | 738 | |||
AK093589.1 | - | 6 | 889 | |||
AK127539.1 | - | 5 | 685 | |||
AK303699.1 | - | 5 | 555 | |||
AK312999.1 | - | 6 | 690 | |||
AY704670.1 | - | 6 | 738 | |||
AY704671.1 | - | 6 | 738 | |||
AY704672.1 | - | 6 | 684 | |||
BC036059.1 | - | 6 | 834 | 5298499 | ||
KU257674.1 | - | 6 | 596 |