Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NFKBIA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020529.2 | NP_065390.1 | 3, 4 | 649 | ||
GenBank mRNA | AK311515.1 | - | 2, 3 | 411 | ||
AK313421.1 | - | 3, 4 | 645 | |||
AY033600.1 | - | 3, 4 | 553 | |||
BC002601.2 | - | 3, 4 | 641 | 3161686 | ||
BC004983.1 | - | 3, 4 | 632 | 2957970 | ||
BT007091.1 | - | 3, 4 | 539 | |||
M69043.1 | - | 3, 4 | 633 |