Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acadm | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007382.5 | NP_031408.1 | 12 | 1495 | ||
GenBank mRNA | AK079930.1 | - | 4 | 539 | ||
AK146444.1 | - | 12 | 1300 | |||
BC013498.1 | - | 12 | 1280 | 4222450 | ||
U07159.1 | - | 12 | 1284 |