Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Adam9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270996.1 | NP_001257925.1 | 17 | 2038 | ||
NM_007404.2 | NP_031430.2 | 17 | 2038 | |||
GenBank mRNA | AK122188.1 | - | 17 | 2021 | ||
AK148174.1 | - | 17 | 1924 | |||
AK155582.1 | - | 17 | 2000 | |||
AK155672.1 | - | 17 | 1931 | |||
AK161778.1 | - | 17 | 1961 | |||
BC047156.1 | - | 17 | 1916 | 6488707 | ||
BC052710.1 | - | 17 | 1910 | |||
U41765.1 | - | 17 | 1910 |