Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arntl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007489.4 | NP_031515.1 | 6, 7 | 707 | ||
GenBank mRNA | AB012601.1 | - | 3, 4 | 208 | ||
AB012602.1 | - | 2, 3 | 187 | |||
AB014494.1 | - | 6, 7 | 593 | |||
AB015203.1 | - | 2, 3 | 187 | |||
AK143267.1 | - | 6, 7 | 635 | |||
AK146628.1 | - | 6, 7 | 707 | |||
AK147145.1 | - | 6, 7 | 707 | |||
AY690602.1 | - | 3, 4 | 208 | |||
BC025973.1 | - | 4, 5 | 458 | 3980719 |