Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Slc7a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301424.1 | NP_001288353.1 | 4, 5 | 701 | ||
NM_007513.4 | NP_031539.3 | 4, 5 | 817 | |||
GenBank mRNA | AK139342.1 | - | 4, 5 | 793 | ||
AK147991.1 | - | 4, 5 | 726 | |||
AK163202.1 | - | 4, 5 | 701 | |||
BC068234.1 | - | 4, 5 | 709 | |||
BC145779.1 | - | 3, 4 | 622 | 40131082 | ||
BC145781.1 | - | 3, 4 | 622 | 40131084 | ||
JN226407.1 | - | 2, 3 | 537 | |||
JN226408.1 | - | 2, 3 | 537 | |||
JN226410.1 | - | 2, 3 | 537 | |||
M26687.1 | - | 4, 5 | 735 |