Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnrnpd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077265.2 | NP_001070733.1 | 1, 2 | 544 | ||
NM_007516.3 | NP_031542.2 | 1, 2 | 544 | |||
GenBank mRNA | AK077409.1 | - | 1, 2 | 533 | ||
AK135733.1 | - | 1, 2 | 490 | |||
BC011172.1 | - | 1, 2 | 219 | |||
BC049098.1 | - | 1, 2 | 544 | 5717286 |