Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Bdnf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001048139.1 | NP_001041604.1 | 2 | 762 | ||
NM_001048141.1 | NP_001041606.1 | 2 | 602 | |||
NM_001048142.1 | NP_001041607.1 | 2 | 610 | |||
NM_001285416.1 | NP_001272345.1 | 2 | 679 | |||
NM_001285417.1 | NP_001272346.1 | 2 | 476 | |||
NM_001285418.1 | NP_001272347.1 | 2 | 467 | |||
NM_001285419.1 | NP_001272348.1 | 2 | 456 | |||
NM_001285420.1 | NP_001272349.1 | 2 | 344 | |||
NM_001285421.1 | NP_001272350.1 | 2 | 542 | |||
NM_001285422.1 | NP_001272351.1 | 1 | 724 | |||
NM_007540.4 | NP_031566.4 | 2 | 903 | |||
GenBank mRNA | AK017559.1 | - | 2 | 544 | ||
AK033127.1 | - | 2 | 556 | |||
AY057908.1 | - | 2 | 903 | |||
AY057909.1 | - | 2 | 467 | |||
AY057910.1 | - | 2 | 679 | |||
AY057911.1 | - | 2 | 762 | |||
AY057912.1 | - | 2 | 394 | |||
AY057913.1 | - | 2 | 602 | |||
AY057914.1 | - | 2 | 610 | |||
AY231131.1 | - | 2 | 416 | |||
AY231132.1 | - | 3 | 491 | |||
BC034862.1 | - | 1 | 269 | 1397218 | ||
EF125669.1 | - | 2 | 903 | |||
EF125670.1 | - | 2 | 468 | |||
EF125671.1 | - | 2 | 680 | |||
EF125672.1 | - | 2 | 764 | |||
EF125673.1 | - | 2 | 602 | |||
EF125674.1 | - | 2 | 610 | |||
EF125681.1 | - | 2 | 456 | |||
EF125682.1 | - | 2 | 344 | |||
EF125683.1 | - | 2 | 476 | |||
EF125684.1 | - | 2 | 542 | |||
EF125685.1 | - | 1 | 721 | |||
X55573.1 | - | 2 | 319 |