Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rhod | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007485.4 | NP_031511.1 | 4 | 441 | ||
GenBank mRNA | AK008880.1 | - | 4 | 442 | ||
BC005652.1 | - | 4 | 438 | |||
BC047989.1 | - | 4 | 450 | 6333308 | ||
D89821.1 | - | 452 | ||||
X84325.1 | - | 4 | 368 |