Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aup1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301649.1 | NP_001288578.1 | 1487 | |||
NM_007517.4 | NP_031543.2 | 1638 | ||||
GenBank mRNA | BC012699.1 | - | 1271 | 3986517 | ||
BC016485.1 | - | 1431 | 3962982 | |||
U41736.1 | - | 1436 |