Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdh6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007666.3 | NP_031692.2 | 3 | 510 | ||
GenBank mRNA | AK134611.1 | - | 3 | 547 | ||
BC107008.1 | - | 2 | 251 | 40047565 | ||
BC107013.1 | - | 2 | 251 | 40048308 | ||
CN704843.1 | - | 3 | 510 | |||
D82029.1 | - | 2 | 251 |