Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Canx | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110499.1 | NP_001103969.1 | 2, 3 | 366 | ||
NM_001110500.1 | NP_001103970.1 | 2, 3 | 334 | |||
NM_007597.3 | NP_031623.1 | 2, 3 | 370 | |||
GenBank mRNA | AK028279.1 | - | 2, 3 | 297 | ||
AK084175.1 | - | 2, 3 | 310 | |||
AK159298.1 | - | 2, 3 | 354 | |||
AK165508.1 | - | 2, 3 | 269 | |||
AK166649.1 | - | 2, 3 | 347 | |||
BC012408.1 | - | 2, 3 | 288 | 3497769 | ||
BC040244.1 | - | 2, 3 | 297 | 2582119 | ||
L18888.1 | - | 1, 2 | 182 | |||
L23865.1 | - | 1, 2 | 122 |