Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301067.1 | NP_001287996.1 | 5 | 629 | ||
NM_007598.4 | NP_031624.2 | 5 | 613 | |||
GenBank mRNA | AK053351.1 | - | 5 | 437 | ||
AK128889.1 | - | 5 | 435 | |||
AK137222.1 | - | 5 | 418 | |||
AK144094.1 | - | 5 | 421 | |||
AK154002.1 | - | 5 | 633 | |||
AK156556.1 | - | 5 | 447 | |||
AK158150.1 | - | 4 | 1277 | |||
AK165565.1 | - | 5 | 416 | |||
AK169652.1 | - | 5 | 408 | |||
AK170901.1 | - | 5 | 424 | |||
AK171388.1 | - | 5 | 431 | |||
BC005446.1 | - | 4 | 412 | 3600514 | ||
BC005472.1 | - | 4 | 405 | 3590890 | ||
L12367.1 | - | 5 | 439 |