Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Capn1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001110504.1 | NP_001103974.1 | 16, 17 | 1811 | ||
NM_007600.3 | NP_031626.1 | 16, 17 | 1863 | |||
GenBank mRNA | AF021847.1 | - | 16, 17 | 1742 | ||
AF084459.1 | - | 15, 16 | 1729 | |||
AK088547.1 | - | 16, 17 | 1783 | |||
AK149090.1 | - | 12, 13 | 1211 | |||
AK149706.1 | - | 16, 17 | 1804 | |||
AK150084.1 | - | 16, 17 | 1802 | |||
AK160940.1 | - | 16, 17 | 1803 | |||
AK165258.1 | - | 16, 17 | 1823 | |||
AK168063.1 | - | 16, 17 | 1802 | |||
AK171405.1 | - | 16, 17 | 1766 | |||
BC026138.1 | - | 16, 17 | 1778 | 4218148 | ||
BC050276.1 | - | 16, 17 | 1779 | 4923347 |