Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 5 | 302 | ||
GenBank mRNA | AK031577.1 | - | 5 | 298 | ||
AK132496.1 | - | 5 | 298 | |||
AK157724.1 | - | 4 | 947 | |||
AK159212.1 | - | 5 | 303 | |||
AK159408.1 | - | 5 | 299 | |||
AK159578.1 | - | 5 | 299 | |||
AK160098.1 | - | 5 | 302 | |||
AK160787.1 | - | 5 | 298 | |||
AK160788.1 | - | 5 | 297 | |||
AK160790.1 | - | 5 | 298 | |||
AK160946.1 | - | 5 | 301 | |||
AK161017.1 | - | 5 | 303 | |||
AK161605.1 | - | 5 | 298 | |||
AK164808.1 | - | 5 | 298 | |||
BC007158.1 | - | 5 | 282 | 3487477 | ||
BC042503.1 | - | 5 | 290 | 3586272 | ||
X58251.1 | - | 5 | 238 |