Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cux2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007804.2 | NP_031830.2 | 7, 8 | 833 | ||
GenBank mRNA | BC065113.1 | - | 7, 8 | 833 | 30532644 | |
U45665.1 | - | 7, 8 | 654 |