Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col19a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007733.2 | NP_031759.2 | 3, 4 | 289 | ||
GenBank mRNA | AB000636.1 | - | 3, 4 | 263 | ||
AK149136.1 | - | 3, 4 | 282 | |||
BC118970.2 | - | 3, 4 | 242 | 40097873 | ||
BY247446.1 | - | 3, 4 | 289 |