Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cpeb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252525.1 | NP_001239454.1 | 2 | 287 | ||
NM_001252526.1 | NP_001239455.1 | 2 | 287 | |||
NM_007755.5 | NP_031781.1 | 2 | 287 | |||
GenBank mRNA | AK136088.1 | - | 2 | 290 | ||
BC125476.1 | - | 2 | 204 | 40129991 | ||
BC144948.1 | - | 2 | 204 | |||
BY720025.1 | - | 2 | 290 | |||
Y08260.1 | - | 2 | 206 |