Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csn1s1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286015.1 | NP_001272944.1 | 9, 10 | 336 | ||
NM_001286016.1 | NP_001272945.1 | 10, 11 | 360 | |||
NM_007784.3 | NP_031810.1 | 10, 11 | 360 | |||
GenBank mRNA | AK052808.1 | - | 10, 11 | 358 | ||
AK085690.1 | - | 10, 11 | 361 | |||
BC002101.1 | - | 10, 11 | 329 | 3484624 | ||
BC003859.1 | - | 10, 11 | 344 | 3486384 | ||
BC006024.1 | - | 9, 10 | 320 | 3592625 | ||
BC007192.1 | - | 10, 11 | 344 | |||
BC008278.1 | - | 10, 11 | 344 | 2812523 | ||
BC040246.1 | - | 10, 11 | 356 | 2647887 | ||
M36780.1 | - | 9, 10 | 299 |