Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Csf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001113529.1 | NP_001107001.1 | 9 | 1213 | ||
NM_007778.4 | NP_031804.3 | 9 | 2098 | |||
GenBank mRNA | AK138489.1 | - | 9 | 1872 | ||
AK154261.1 | - | 9 | 2099 | |||
AK160995.1 | - | 9 | 1872 | |||
AK166370.1 | - | 9 | 1871 | |||
AK170154.1 | - | 9 | 1808 | |||
BC025593.1 | - | 9 | 999 | |||
BC066187.1 | - | 9 | 1852 | 30106218 | ||
BC066200.1 | - | 9 | 1852 | 30100524 | ||
BC066205.1 | - | 9 | 1852 | 30100601 | ||
M21952.1 | - | 9 | 1851 | |||
S78392.1 | - | 9 | 966 |